Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAATATATATGCGAATACCTTAT[C/G]TTCAACACTTTCTTTAATGTGTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611965 | ||||||||||||||||||||
Literature Links: |
C3orf49 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C3orf49 - chromosome 3 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THOC7 - THO complex 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285387.2 | 554 | Missense Mutation | CAT,GAT | H,D 106 | NP_001272316.1 | |
NM_001285404.1 | 554 | Missense Mutation | CAT,GAT | H,D 106 | NP_001272333.1 | |
NM_025075.3 | 554 | Missense Mutation | CAT,GAT | H,D 158 | NP_079351.2 | |
XM_006713339.3 | 554 | Missense Mutation | CAT,GAT | H,D 106 | XP_006713402.1 |