Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCCATGTTCACTAGCGGCCTTA[C/T]AGAAAGTACTCAAAAAGAAGTTCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616607 | ||||||||||||||||||||
Literature Links: |
KBTBD8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KBTBD8 - kelch repeat and BTB domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032505.2 | 89 | Missense Mutation | ACA,ATA | T,I 84 | NP_115894.2 | |
XM_005264771.3 | 89 | Missense Mutation | ACA,ATA | T,I 7 | XP_005264828.1 |