Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146691 MIM: 615150 MIM: 612911 MIM: 606031 | ||||||||||||||||||||
Literature Links: |
DALRD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DALRD3 - DALR anticodon binding domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IMPDH2 - inosine monophosphate dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR191 - microRNA 191 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR425 - microRNA 425 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFAF3 - NADH:ubiquinone oxidoreductase complex assembly factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199069.1 | 457 | Silent Mutation | ACA,ACG | T,T 146 | NP_951032.1 | |
NM_199070.1 | 457 | Silent Mutation | ACA,ACG | T,T 89 | NP_951033.1 | |
NM_199073.1 | 457 | Silent Mutation | ACA,ACG | T,T 89 | NP_951047.1 | |
NM_199074.1 | 457 | Silent Mutation | ACA,ACG | T,T 89 | NP_951056.1 |
QRICH1 - glutamine rich 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR6 - WD repeat domain 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |