Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGCTGCTGCTGCTGGACTATGC[C/T]GCTAACCGCCTGCTGGCCTGTGGCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601055 | |||||||||||||||||||||||
Literature Links: |
PLXNA1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLXNA1 - plexin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032242.3 | 1064 | Silent Mutation | GCC,GCT | A,A 123 | NP_115618.3 | |
XM_011512908.2 | 1064 | Silent Mutation | GCC,GCT | A,A 141 | XP_011511210.1 | |
XM_011512909.1 | 1064 | Silent Mutation | GCC,GCT | A,A 141 | XP_011511211.1 |