Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGCTCTCCTCAACATGAGAGCT[A/G]CACCCCTCCTCCTGGCCAGGGCAGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 171790 | |||||||||||||||||||||||
Literature Links: |
ACPP PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ACPP - acid phosphatase, prostate | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099.4 | 64 | Missense Mutation | ACA,GCA | T,A 4 | NP_001090.2 | |
NM_001134194.1 | 64 | Missense Mutation | ACA,GCA | T,A 4 | NP_001127666.1 | |
NM_001292037.1 | 64 | Missense Mutation | ACA,GCA | T,A 4 | NP_001278966.1 | |
XM_011512946.1 | 64 | Missense Mutation | ACA,GCA | T,A 4 | XP_011511248.1 | |
XM_011512947.2 | 64 | Missense Mutation | ACA,GCA | T,A 4 | XP_011511249.1 |