Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCGTGCTGCTTGCACGGATACC[C/T]CAGCGGGCGCCACGGTGGCTCCCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610298 | |||||||||||||||||||||||
Literature Links: |
ABHD10 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ABHD10 - abhydrolase domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272069.1 | 303 | Silent Mutation | CCC,CCT | P,P 39 | NP_001258998.1 | |
NM_018394.3 | 303 | Silent Mutation | CCC,CCT | P,P 39 | NP_060864.1 | |
XM_011512960.2 | 303 | Silent Mutation | CCC,CCT | P,P 39 | XP_011511262.1 |
PHLDB2 - pleckstrin homology like domain family B member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |