Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTTGGGGGGCTGCCCTTCCTCC[A/G]TAGCATCGAGGAGCTGCTGCAGGTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612120 | |||||||||||||||||||||||
Literature Links: |
CIDEC PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CIDEC - cell death inducing DFFA like effector c | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199551.1 | 733 | Missense Mutation | ACG,ATG | T,M 226 | NP_001186480.1 | |
NM_001199552.1 | 733 | Missense Mutation | ACG,ATG | T,M 216 | NP_001186481.1 | |
NM_001199623.1 | 733 | Missense Mutation | ACG,ATG | T,M 229 | NP_001186552.1 | |
NM_001321142.1 | 733 | Missense Mutation | ACG,ATG | T,M 216 | NP_001308071.1 | |
NM_001321143.1 | 733 | Missense Mutation | ACG,ATG | T,M 142 | NP_001308072.1 | |
NM_001321144.1 | 733 | Missense Mutation | ACG,ATG | T,M 142 | NP_001308073.1 | |
NM_022094.3 | 733 | Missense Mutation | ACG,ATG | T,M 216 | NP_071377.2 |