Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTGACACAGCAGTGCGTGTAGG[C/T]GATCACCTCCGTCACTTGCACAGCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601159 MIM: 601268 | |||||||||||||||||||||||
Literature Links: |
CCR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCR1 - C-C motif chemokine receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001295.2 | 939 | Missense Mutation | ACC,GCC | T,A 290 | NP_001286.1 |
CCR3 - C-C motif chemokine receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |