Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCTCTACCGAACTCCAACCATC[A/G]TTGCCTTGGTGGTCTTGCTTGTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603161 | ||||||||||||||||||||
Literature Links: |
ENTPD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENTPD3 - ectonucleoside triphosphate diphosphohydrolase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001248.3 | 191 | Missense Mutation | ATT,GTT | I,V 25 | NP_001239.2 | |
NM_001291960.1 | 191 | Missense Mutation | ATT,GTT | I,V 25 | NP_001278889.1 | |
NM_001291961.1 | 191 | Missense Mutation | ATT,GTT | I,V 25 | NP_001278890.1 | |
XM_011534266.2 | 191 | Missense Mutation | ATT,GTT | I,V 25 | XP_011532568.1 |
ENTPD3-AS1 - ENTPD3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |