Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGGCAAAGCACAACTTTCCTCC[A/G]CTTCTCTCCTCGGAGCCGGGCTCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607147 | |||||||||||||||||||||||
Literature Links: |
NECTIN3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NECTIN3 - nectin cell adhesion molecule 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243286.1 | 318 | Missense Mutation | ACT,GCT | T,A 22 | NP_001230215.1 | |
NM_001243288.1 | 318 | Intron | NP_001230217.1 | |||
NM_015480.2 | 318 | Missense Mutation | ACT,GCT | T,A 22 | NP_056295.1 | |
XM_011512663.1 | 318 | Missense Mutation | ACT,GCT | T,A 22 | XP_011510965.1 | |
XM_017006123.1 | 318 | Missense Mutation | ACT,GCT | T,A 22 | XP_016861612.1 | |
XM_017006124.1 | 318 | Intron | XP_016861613.1 | |||
XM_017006125.1 | 318 | Intron | XP_016861614.1 | |||
XM_017006126.1 | 318 | Missense Mutation | ACT,GCT | T,A 22 | XP_016861615.1 | |
XM_017006127.1 | 318 | Intron | XP_016861616.1 |
PVRL3-AS1 - PVRL3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |