Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCTTTCTCTGGGAGAAGGGTTC[A/G]AATCCCAGAAGTCTGAACACTGCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601898 | |||||||||||||||||||||||
Literature Links: |
GHSR PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GHSR - growth hormone secretagogue receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004122.2 | 1063 | Intron | NP_004113.1 | |||
NM_198407.2 | 1063 | Silent Mutation | TTC,TTT | F,F 340 | NP_940799.1 |