Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTTGGGGATCCAGAGCCATGTC[A/G]GACCTGCTACTACTGGGCCTGATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RAD54L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAD54L2 - RAD54-like 2 (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX264 - testis expressed 264 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129884.2 | 314 | Silent Mutation | TCA,TCG | S,S 2 | NP_001123356.1 | |
NM_001243725.1 | 314 | Silent Mutation | TCA,TCG | S,S 2 | NP_001230654.1 | |
NM_001243726.1 | 314 | Silent Mutation | TCA,TCG | S,S 2 | NP_001230655.1 | |
NM_001243727.2 | 314 | Silent Mutation | TCA,TCG | S,S 2 | NP_001230656.1 | |
NM_001278195.1 | 314 | Silent Mutation | TCA,TCG | S,S 2 | NP_001265124.1 | |
NM_015926.5 | 314 | Silent Mutation | TCA,TCG | S,S 2 | NP_057010.1 | |
XM_006713195.3 | 314 | Silent Mutation | TCA,TCG | S,S 2 | XP_006713258.1 | |
XM_006713197.3 | 314 | Silent Mutation | TCA,TCG | S,S 2 | XP_006713260.1 | |
XM_006713198.2 | 314 | UTR 5 | XP_006713261.1 | |||
XM_011533805.2 | 314 | Silent Mutation | TCA,TCG | S,S 2 | XP_011532107.1 | |
XM_017006573.1 | 314 | Silent Mutation | TCA,TCG | S,S 41 | XP_016862062.1 | |
XM_017006574.1 | 314 | Silent Mutation | TCA,TCG | S,S 2 | XP_016862063.1 |