Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACATGTGAGAGTGTGTGAGTCT[C/T]TATCCATAGGCAGTGCTTAAACTCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601325 | |||||||||||||||||||||||
Literature Links: |
CNTN3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CNTN3 - contactin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020872.2 | 2076 | UTR 3 | NP_065923.1 | |||
XM_005264757.3 | 2076 | UTR 3 | XP_005264814.1 | |||
XM_011533768.2 | 2076 | UTR 3 | XP_011532070.1 | |||
XM_017006507.1 | 2076 | UTR 3 | XP_016861996.1 | |||
XM_017006508.1 | 2076 | UTR 3 | XP_016861997.1 | |||
XM_017006509.1 | 2076 | UTR 3 | XP_016861998.1 |