Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGTTTGGTCACTTTGGAGCATA[C/T]CCTCTCGATGACCATGGGATTTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604419 | ||||||||||||||||||||
Literature Links: |
POLQ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLQ - DNA polymerase theta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199420.3 | 7649 | Missense Mutation | GAT,GGT | D,G 2507 | NP_955452.3 | |
XM_011512343.2 | 7649 | Missense Mutation | GAT,GGT | D,G 2461 | XP_011510645.1 | |
XM_011512347.2 | 7649 | Missense Mutation | GAT,GGT | D,G 2507 | XP_011510649.1 | |
XM_011512348.2 | 7649 | Missense Mutation | GAT,GGT | D,G 2336 | XP_011510650.1 | |
XM_011512352.2 | 7649 | Intron | XP_011510654.1 | |||
XM_011512354.2 | 7649 | Missense Mutation | GAT,GGT | D,G 1744 | XP_011510656.1 | |
XM_017005565.1 | 7649 | Missense Mutation | GAT,GGT | D,G 2117 | XP_016861054.1 |