Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTCTGACAAGAAGTTTTGTCCT[C/T]CCCAAGGCATATGGCATCAAGGCTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 102645 MIM: 142408 MIM: 614472 | |||||||||||||||||||||||
Literature Links: |
APEH PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
APEH - acylaminoacyl-peptide hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001640.3 | 3073 | Intron | NP_001631.3 | |||
XM_005265097.2 | 3073 | Intron | XP_005265154.1 | |||
XM_005265098.4 | 3073 | Intron | XP_005265155.1 | |||
XM_011533658.2 | 3073 | Intron | XP_011531960.1 | |||
XM_011533659.1 | 3073 | Intron | XP_011531961.1 | |||
XM_011533660.2 | 3073 | Intron | XP_011531962.1 | |||
XM_011533661.2 | 3073 | Intron | XP_011531963.1 | |||
XM_011533662.2 | 3073 | Intron | XP_011531964.1 | |||
XM_011533663.2 | 3073 | Intron | XP_011531965.1 | |||
XM_017006285.1 | 3073 | Intron | XP_016861774.1 | |||
XM_017006286.1 | 3073 | Intron | XP_016861775.1 |
MST1 - macrophage stimulating 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020998.3 | 3073 | UTR 3 | NP_066278.3 | |||
XM_006713166.1 | 3073 | Intron | XP_006713229.1 | |||
XM_011533732.1 | 3073 | Intron | XP_011532034.1 | |||
XM_011533737.2 | 3073 | Intron | XP_011532039.1 | |||
XM_011533738.2 | 3073 | Intron | XP_011532040.1 | |||
XM_017006460.1 | 3073 | Intron | XP_016861949.1 | |||
XM_017006461.1 | 3073 | Intron | XP_016861950.1 | |||
XM_017006462.1 | 3073 | UTR 3 | XP_016861951.1 | |||
XM_017006463.1 | 3073 | Intron | XP_016861952.1 | |||
XM_017006464.1 | 3073 | UTR 3 | XP_016861953.1 | |||
XM_017006465.1 | 3073 | Intron | XP_016861954.1 | |||
XM_017006466.1 | 3073 | Intron | XP_016861955.1 | |||
XM_017006467.1 | 3073 | Intron | XP_016861956.1 | |||
XM_017006468.1 | 3073 | Intron | XP_016861957.1 |
RNF123 - ring finger protein 123 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |