Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATGTTTAAAGTCTTCAGTTGT[A/G]GTAATTCTTCCCACTTTGCAGTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614169 MIM: 612778 | ||||||||||||||||||||
Literature Links: |
NBEAL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NBEAL2 - neurobeachin like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRADDP - neurotrophin receptor associated death domain, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD2 - SET domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014159.6 | 9049 | Silent Mutation | ACC,ACT | T,T 2500 | NP_054878.5 | |
XM_011533631.2 | 9049 | Silent Mutation | ACC,ACT | T,T 2526 | XP_011531933.1 | |
XM_011533632.2 | 9049 | Silent Mutation | ACC,ACT | T,T 2508 | XP_011531934.1 | |
XM_011533633.2 | 9049 | Silent Mutation | ACC,ACT | T,T 2481 | XP_011531935.1 | |
XM_011533634.1 | 9049 | Silent Mutation | ACC,ACT | T,T 2456 | XP_011531936.1 | |
XM_017006270.1 | 9049 | Silent Mutation | ACC,ACT | T,T 2426 | XP_016861759.1 |