Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGCGACGGCGGCCTACGAGCAG[C/T]TGAAGCTGTGAGTCCCACGGGCCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604543 MIM: 606569 | |||||||||||||||||||||||
Literature Links: |
LIMD1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LIMD1 - LIM domains containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIMD1-AS1 - LIMD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SACM1L - SAC1 suppressor of actin mutations 1-like (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319072.1 | 250 | UTR 5 | NP_001306001.1 | |||
NM_001319073.1 | 250 | UTR 5 | NP_001306002.1 | |||
NM_014016.4 | 250 | Silent Mutation | CTG,TTG | L,L 9 | NP_054735.3 | |
XM_011533500.2 | 250 | UTR 5 | XP_011531802.1 |