Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAACCTTGGTCTTGGGCTTAACA[C/G]GCAATGGGCGGATCATGACTGTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605958 | ||||||||||||||||||||
Literature Links: |
TRAIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRAIP - TRAF interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005879.2 | 1150 | Missense Mutation | CCT,CGT | P,R 439 | NP_005870.2 | |
XM_011533264.1 | 1150 | Missense Mutation | CCT,CGT | P,R 362 | XP_011531566.1 | |
XM_017005526.1 | 1150 | Missense Mutation | CCT,CGT | P,R 340 | XP_016861015.1 |