Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACGAGCAGCATCACCCTCAGGG[A/G]CTGAGGTAGCTCCACTCGCAGCTGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614993 MIM: 605958 | |||||||||||||||||||||||
Literature Links: |
CAMKV PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CAMKV - CaM kinase like vesicle associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320147.1 | 1573 | Intron | NP_001307076.1 | |||
NM_024046.4 | 1573 | Missense Mutation | GCC,GTC | A,V 365 | NP_076951.2 |
TRAIP - TRAF interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |