Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCACAGGGGCTCCTGGATCATC[A/G]GGTGAGCCCTTTATTCCACAGACTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600249 MIM: 610803 | |||||||||||||||||||||||
Literature Links: |
ALDH1L1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ALDH1L1 - aldehyde dehydrogenase 1 family member L1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270364.1 | Intron | NP_001257293.1 | ||||
NM_001270365.1 | Intron | NP_001257294.1 | ||||
NM_012190.3 | Intron | NP_036322.2 | ||||
XM_006713481.2 | Intron | XP_006713544.1 | ||||
XM_011512355.1 | Intron | XP_011510657.1 | ||||
XM_017005613.1 | Intron | XP_016861102.1 | ||||
XM_017005614.1 | Intron | XP_016861103.1 |
ALDH1L1-AS1 - ALDH1L1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC41A3 - solute carrier family 41 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |