Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAGGGTACCAGACCAAGTGAAT[A/G]CGACAGGGAATTATTTCCTAGGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606639 MIM: 605090 | ||||||||||||||||||||
Literature Links: |
GFM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GFM1 - G elongation factor mitochondrial 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RARRES1 - retinoic acid receptor responder 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002888.3 | 1240 | Intron | NP_002879.2 | |||
NM_206963.1 | 1240 | Silent Mutation | CGC,CGT | R,R 251 | NP_996846.1 | |
XM_005247686.4 | 1240 | Silent Mutation | CGC,CGT | R,R 393 | XP_005247743.3 |