Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGACCAAAATACTGCAGCACCA[A/C]GAGGAGAGTGGTATAAGACAACCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 162641 | ||||||||||||||||||||
Literature Links: |
NPY1R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPY1R - neuropeptide Y receptor Y1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000909.5 | 2546 | Missense Mutation | GTG,TTG | V,L 216 | NP_000900.1 | |
XM_005263031.3 | 2546 | Missense Mutation | GTG,TTG | V,L 216 | XP_005263088.1 | |
XM_011532010.2 | 2546 | Missense Mutation | GTG,TTG | V,L 216 | XP_011530312.1 |