Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGCAGCTCTGTGCCCGACTGAG[C/G]GACCTTCTGACCGACTTTGTGCAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC105377623 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105377623 - uncharacterized LOC105377623 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP38 - ubiquitin specific peptidase 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290325.1 | 978 | Missense Mutation | AGC,AGG | S,R 148 | NP_001277254.1 | |
NM_001290326.1 | 978 | UTR 5 | NP_001277255.1 | |||
NM_032557.6 | 978 | Missense Mutation | AGC,AGG | S,R 148 | NP_115946.2 | |
XM_011532360.2 | 978 | Missense Mutation | AGC,AGG | S,R 148 | XP_011530662.1 |