Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAATCAGCTTGAGTGTAAACGT[A/G]GCTTCTGCCACATTATGAAAGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZGRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZGRF1 - zinc finger GRF-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018392.4 | 5974 | Silent Mutation | GCC,GCT | A,A 1938 | NP_060862.3 | |
XM_005263115.3 | 5974 | Silent Mutation | GCC,GCT | A,A 1938 | XP_005263172.1 | |
XM_011532091.2 | 5974 | Silent Mutation | GCC,GCT | A,A 1921 | XP_011530393.1 | |
XM_011532092.2 | 5974 | Silent Mutation | GCC,GCT | A,A 1918 | XP_011530394.1 | |
XM_011532093.2 | 5974 | Silent Mutation | GCC,GCT | A,A 1880 | XP_011530395.1 | |
XM_011532094.2 | 5974 | Silent Mutation | GCC,GCT | A,A 1852 | XP_011530396.1 | |
XM_011532096.2 | 5974 | Intron | XP_011530398.1 | |||
XM_011532097.2 | 5974 | Silent Mutation | GCC,GCT | A,A 987 | XP_011530399.1 | |
XM_011532098.2 | 5974 | Silent Mutation | GCC,GCT | A,A 895 | XP_011530400.1 | |
XM_011532099.2 | 5974 | Silent Mutation | GCC,GCT | A,A 895 | XP_011530401.1 | |
XM_017008369.1 | 5974 | Silent Mutation | GCC,GCT | A,A 1918 | XP_016863858.1 | |
XM_017008370.1 | 5974 | Silent Mutation | GCC,GCT | A,A 1863 | XP_016863859.1 | |
XM_017008371.1 | 5974 | Silent Mutation | GCC,GCT | A,A 1843 | XP_016863860.1 | |
XM_017008372.1 | 5974 | Silent Mutation | GCC,GCT | A,A 688 | XP_016863861.1 | |
XM_017008373.1 | 5974 | Silent Mutation | GCC,GCT | A,A 688 | XP_016863862.1 |