Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTTTTCATATTTTGATTTAGGA[C/T]TGTTAAAATGAGTCTTCGGAAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607286 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSM6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LSM6 - LSM6 homolog, U6 small nuclear RNA and mRNA degradation associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007080.2 | 107 | UTR 5 | NP_009011.1 | |||
XM_017007672.1 | 107 | UTR 5 | XP_016863161.1 | |||
XM_017007673.1 | 107 | UTR 5 | XP_016863162.1 |