Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACCTGGATTTTTATTTCAATTT[A/G]TACCTTATATGAAAAAATACAAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607545 | ||||||||||||||||||||
Literature Links: |
MSMO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MSMO1 - methylsterol monooxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017369.2 | 353 | Intron | NP_001017369.1 | |||
NM_006745.4 | 353 | Missense Mutation | ATA,GTA | I,V 75 | NP_006736.1 | |
XM_005263176.2 | 353 | Missense Mutation | ATA,GTA | I,V 75 | XP_005263233.1 |