Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATACCTAAGGAATTTATAATGA[A/C]ATACAAACGTGGAGAGATAAATCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614130 | ||||||||||||||||||||
Literature Links: |
ADAD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAD1 - adenosine deaminase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159285.1 | 519 | Missense Mutation | AAA,ACA | K,T 89 | NP_001152757.1 | |
NM_001159295.1 | 519 | Missense Mutation | AAA,ACA | K,T 71 | NP_001152767.1 | |
NM_139243.3 | 519 | Missense Mutation | AAA,ACA | K,T 89 | NP_640336.1 | |
XM_005262741.2 | 519 | Missense Mutation | AAA,ACA | K,T 89 | XP_005262798.1 | |
XM_005262743.2 | 519 | Missense Mutation | AAA,ACA | K,T 89 | XP_005262800.1 | |
XM_005262744.2 | 519 | Missense Mutation | AAA,ACA | K,T 89 | XP_005262801.1 | |
XM_005262745.2 | 519 | Missense Mutation | AAA,ACA | K,T 89 | XP_005262802.1 | |
XM_011531603.1 | 519 | Missense Mutation | AAA,ACA | K,T 89 | XP_011529905.1 |