Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGGGCAGGGGCTCCCGTTGTTGG[C/T]GGGCTGGACCCGGACGTAGCGAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605918 | ||||||||||||||||||||
Literature Links: |
SPON2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPON2 - spondin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128325.2 | 1257 | Missense Mutation | ACC,GCC | T,A 310 | NP_001121797.1 | |
NM_001199021.1 | 1257 | Missense Mutation | ACC,GCC | T,A 310 | NP_001185950.1 | |
NM_012445.3 | 1257 | Missense Mutation | ACC,GCC | T,A 310 | NP_036577.1 |