Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTCTATTCCAATCTGAGTGGCT[A/G]TAAACACCCATGTAGCAACACAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX7B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX7B2 - cytochrome c oxidase subunit 7B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130902.2 | 239 | Missense Mutation | ACA,ATA | T,I 57 | NP_570972.2 | |
XM_005248056.4 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_005248113.1 | |
XM_011513630.2 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_011511932.1 | |
XM_011513631.2 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_011511933.1 | |
XM_011513632.2 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_011511934.1 | |
XM_011513633.2 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_011511935.1 | |
XM_011513634.2 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_011511936.1 | |
XM_011513635.2 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_011511937.1 | |
XM_011513636.1 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_011511938.1 | |
XM_011513637.2 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_011511939.1 | |
XM_011513638.2 | 239 | Missense Mutation | ACA,ATA | T,I 57 | XP_011511940.1 |