Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAGAATGGAAGTTTCAAGGAAG[C/T]TTTGAATGTCATCAATACTCACACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 172439 MIM: 602122 | ||||||||||||||||||||
Literature Links: |
PAICS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAICS - phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRP72 - signal recognition particle 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267722.1 | 231 | Missense Mutation | GCT,GTT | A,V 64 | NP_001254651.1 | |
NM_006947.3 | 231 | Missense Mutation | GCT,GTT | A,V 64 | NP_008878.3 | |
XM_005265765.4 | 231 | Missense Mutation | GCT,GTT | A,V 179 | XP_005265822.3 |