Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTTGTGTACTGATCATCCAGAA[C/T]ATACCTAGAGTAAGACACACCATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600583 MIM: 600058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEC - tec protein tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003215.2 | 2837 | Missense Mutation | ATT,GTT | I,V 514 | NP_003206.2 | |
XM_011513737.2 | 2837 | Missense Mutation | ATT,GTT | I,V 478 | XP_011512039.1 | |
XM_011513741.1 | 2837 | UTR 3 | XP_011512043.1 | |||
XM_017008569.1 | 2837 | Missense Mutation | ATT,GTT | I,V 398 | XP_016864058.1 |
TXK - TXK tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |