Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTCCAGGGCCTGGGCCTCTCTG[A/G]CTGAGAAACTAGGAAGTCACTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601519 MIM: 160782 MIM: 180072 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5I PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5I - ATP synthase, H+ transporting, mitochondrial Fo complex subunit E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFSD7 - major facilitator superfamily domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294341.1 | 633 | Intron | NP_001281270.1 | |||
NM_001294342.1 | 633 | Intron | NP_001281271.1 | |||
NM_032219.3 | 633 | Intron | NP_115595.2 | |||
XM_005272297.1 | 633 | Intron | XP_005272354.1 | |||
XM_005272299.4 | 633 | Intron | XP_005272356.1 | |||
XM_006713918.1 | 633 | Intron | XP_006713981.1 | |||
XM_006713919.1 | 633 | Intron | XP_006713982.1 | |||
XM_011513568.1 | 633 | Intron | XP_011511870.1 | |||
XM_011513571.1 | 633 | Intron | XP_011511873.1 | |||
XM_011513572.1 | 633 | Intron | XP_011511874.1 | |||
XM_011513573.2 | 633 | Intron | XP_011511875.1 | |||
XM_011513574.1 | 633 | Intron | XP_011511876.1 | |||
XM_011513575.2 | 633 | Intron | XP_011511877.1 | |||
XM_017008700.1 | 633 | Intron | XP_016864189.1 |
MYL5 - myosin light chain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002477.1 | 633 | Intron | NP_002468.1 | |||
XM_006713886.2 | 633 | Intron | XP_006713949.1 | |||
XM_017008245.1 | 633 | UTR 5 | XP_016863734.1 | |||
XM_017008246.1 | 633 | UTR 5 | XP_016863735.1 | |||
XM_017008247.1 | 633 | UTR 5 | XP_016863736.1 | |||
XM_017008248.1 | 633 | Intron | XP_016863737.1 |
PDE6B - phosphodiesterase 6B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |