Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAAGTGCCGACAGCATTGGTGC[C/T]GTGTTAAATAGCAAAGATGAGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929741 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929741 - uncharacterized LOC101929741 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL14 - methyltransferase like 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020961.3 | 259 | Silent Mutation | GCC,GCT | A,A 33 | NP_066012.1 | |
XM_005263166.4 | 259 | Silent Mutation | GCC,GCT | A,A 33 | XP_005263223.1 | |
XM_011532162.2 | 259 | Silent Mutation | GCC,GCT | A,A 33 | XP_011530464.1 |