Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGGCCAGTTGTTTTCTTGCAAG[A/G]GTGATCTTGGAGATAAAATAAATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607347 MIM: 610851 MIM: 609028 | ||||||||||||||||||||
Literature Links: |
ALPK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALPK1 - alpha kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AP1AR - adaptor related protein complex 1 associated regulatory protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIFA - TRAF interacting protein with forkhead associated domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052864.2 | 725 | Missense Mutation | CCC,CTC | P,L 148 | NP_443096.1 |