Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTGCCAGATTCAGAACTTCCTCA[C/T]GAGCTGGTTCCAAGCCCTTTGGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105377310 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105377310 - syncytin-1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS6P6 - ribosomal protein S6 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM150C - transmembrane protein 150C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080506.1 | 1234 | UTR 3 | NP_001073975.1 |