Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCTGCTCGGAACTGGCCAGCC[A/C]AAACTCACTGGGCCGCCCGGGGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616210 MIM: 231675 | ||||||||||||||||||||
Literature Links: |
C4orf46 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C4orf46 - chromosome 4 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008393.3 | 360 | Missense Mutation | NP_001008394.1 |
ETFDH - electron transfer flavoprotein dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281737.1 | 360 | Intron | NP_001268666.1 | |||
NM_001281738.1 | 360 | Intron | NP_001268667.1 | |||
NM_004453.3 | 360 | Intron | NP_004444.2 |