Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGTTTGGGCTTTTCTCATAAAA[C/T]GTGGCCATCAAAATGTCATAACGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608732 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC39A8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC39A8 - solute carrier family 39 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135146.1 | 2853 | Intron | NP_001128618.1 | |||
NM_001135147.1 | 2853 | UTR 3 | NP_001128619.1 | |||
NM_001135148.1 | 2853 | Intron | NP_001128620.1 | |||
NM_022154.5 | 2853 | Intron | NP_071437.3 | |||
XM_005263177.1 | 2853 | Intron | XP_005263234.1 | |||
XM_011532181.2 | 2853 | Intron | XP_011530483.1 | |||
XM_011532182.1 | 2853 | Intron | XP_011530484.1 | |||
XM_017008540.1 | 2853 | Intron | XP_016864029.1 | |||
XM_017008541.1 | 2853 | Intron | XP_016864030.1 | |||
XM_017008542.1 | 2853 | Intron | XP_016864031.1 |