Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAAAGGTTATGTATATGAATTTA[A/G]AAGTTTCACAGACTGCCAATTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605768 MIM: 603296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAPP1 - dual adaptor of phosphotyrosine and 3-phosphoinositides 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LAMTOR3 - late endosomal/lysosomal adaptor, MAPK and MTOR activator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243736.1 | 3583 | UTR 3 | NP_001230665.1 | |||
NM_021970.3 | 3583 | UTR 3 | NP_068805.1 |