Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGTGGAATTATGTTGCCGGCCT[A/G]TGGCCTTTGTTACTCTAACGGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614138 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RWDD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RWDD4 - RWD domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC11 - trafficking protein particle complex 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021942.5 | 251 | Missense Mutation | ATG,GTG | M,V 17 | NP_068761.4 | |
NM_199053.2 | 251 | Missense Mutation | ATG,GTG | M,V 17 | NP_951008.1 | |
XM_017008537.1 | 251 | Missense Mutation | ATG,GTG | M,V 17 | XP_016864026.1 | |
XM_017008538.1 | 251 | Missense Mutation | ATG,GTG | M,V 17 | XP_016864027.1 | |
XM_017008539.1 | 251 | Intron | XP_016864028.1 |