Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCGTCTCTGAGGAGGCCAAGCAT[C/G]GCACCTTCGTGGGCCGCATCGCGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606307 MIM: 606308 MIM: 606309 MIM: 606310 | ||||||||||||||||||||
Literature Links: |
PCDHA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDHA1 - protocadherin alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018900.3 | 124 | Intron | NP_061723.1 | |||
NM_031410.2 | 124 | Intron | NP_113598.1 | |||
NM_031411.2 | 124 | Intron | NP_113599.1 |
PCDHA2 - protocadherin alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018905.2 | 124 | Intron | NP_061728.1 | |||
NM_031495.1 | 124 | Intron | NP_113683.1 | |||
NM_031496.1 | 124 | Intron | NP_113684.1 |
PCDHA3 - protocadherin alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018906.2 | 124 | Missense Mutation | CGC,GGC | R,G 42 | NP_061729.1 | |
NM_031497.1 | 124 | Missense Mutation | CGC,GGC | R,G 42 | NP_113685.1 |
PCDHA4 - protocadherin alpha 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |