Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCATCTTCAAAGGCCGGATTTAA[C/T]TGTGATTTTGTTGTAGTCTAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609412 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERCC8 - ERCC excision repair 8, CSA ubiquitin ligase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000082.3 | 1128 | Silent Mutation | CAA,CAG | Q,Q 380 | NP_000073.1 | |
NM_001007233.2 | 1128 | Silent Mutation | CAA,CAG | Q,Q 322 | NP_001007234.1 | |
NM_001007234.2 | 1128 | Intron | NP_001007235.1 | |||
NM_001290285.1 | 1128 | Silent Mutation | CAA,CAG | Q,Q 227 | NP_001277214.1 |