Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCGCTGCCTTGGGCCTGCTCAC[C/T]GGGGTGGATGGACCCCCGCCGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610916 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSUN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NSUN2 - NOP2/Sun RNA methyltransferase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193455.1 | 2681 | Missense Mutation | CAG,CGG | Q,R 732 | NP_001180384.1 | |
NM_017755.5 | 2681 | Missense Mutation | CAG,CGG | Q,R 767 | NP_060225.4 |