Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACACAGCCCAGACACAGGCACC[A/G]TGCCTCCGGCAGGTCCACGGGGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604627 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL17B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL17B - interleukin 17B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317987.1 | 731 | Silent Mutation | NP_001304916.1 | |||
NM_014443.2 | 731 | Silent Mutation | NP_055258.1 | |||
XM_011537622.2 | 731 | Missense Mutation | XP_011535924.1 | |||
XM_017009346.1 | 731 | Missense Mutation | XP_016864835.1 | |||
XM_017009347.1 | 731 | Missense Mutation | XP_016864836.1 | |||
XM_017009348.1 | 731 | Missense Mutation | XP_016864837.1 |
PCYOX1L - prenylcysteine oxidase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301054.1 | 731 | Intron | NP_001287983.1 | |||
NM_001301057.1 | 731 | Intron | NP_001287986.1 | |||
NM_024028.3 | 731 | Intron | NP_076933.3 | |||
XM_011537680.1 | 731 | Intron | XP_011535982.1 | |||
XM_011537681.1 | 731 | Intron | XP_011535983.1 | |||
XM_011537682.2 | 731 | Intron | XP_011535984.1 |