Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACATCCAGGGAAGAAAGCACCAT[A/G]AGTCTTTGAGCTTGAAAAGGTGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM267 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM267 - transmembrane protein 267 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022483.4 | 1802 | Missense Mutation | TCA,TTA | S,L 140 | NP_071928.2 | |
XM_005248333.4 | 1802 | Missense Mutation | TCA,TTA | S,L 140 | XP_005248390.1 | |
XM_005248335.3 | 1802 | Missense Mutation | TCA,TTA | S,L 140 | XP_005248392.1 | |
XM_006714486.3 | 1802 | Missense Mutation | TCA,TTA | S,L 140 | XP_006714549.1 | |
XM_011514074.2 | 1802 | Missense Mutation | TCA,TTA | S,L 140 | XP_011512376.1 | |
XM_011514075.2 | 1802 | Missense Mutation | TCA,TTA | S,L 140 | XP_011512377.1 |