Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCTTGTGTGGGTATGAGTGTA[A/G]AGGATGGGGGTATGCCAGGCCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602711 MIM: 603819 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APBB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APBB3 - amyloid beta precursor protein binding family B member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6831 - microRNA 6831 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35A4 - solute carrier family 35 member A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080670.3 | 1008 | Missense Mutation | AAG,GAG | K,E 4 | NP_542401.1 |
SRA1 - steroid receptor RNA activator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |