Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTACCGCTTCGTTCCCTGGATC[A/G]CACTGAACCTAAGCCACAACCCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIER3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIER3 - MIER family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD9 - SET domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171990.2 | 459 | Missense Mutation | ACA,GCA | T,A 25 | NP_001165461.1 | |
NM_001323018.1 | 459 | Intron | NP_001309947.1 | |||
NM_001323019.1 | 459 | Missense Mutation | ACA,GCA | T,A 25 | NP_001309948.1 | |
NM_001323020.1 | 459 | Intron | NP_001309949.1 | |||
NM_001323022.1 | 459 | Intron | NP_001309951.1 | |||
NM_153706.3 | 459 | Missense Mutation | ACA,GCA | T,A 25 | NP_714917.2 | |
XM_005248426.4 | 459 | Intron | XP_005248483.1 | |||
XM_011543139.2 | 459 | Intron | XP_011541441.1 | |||
XM_011543140.2 | 459 | Intron | XP_011541442.1 |