Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGGAGAGTGTCCTAGGATGTC[A/G]TTGGGGCTGGCAGGCAGCCACCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTNL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTNL9 - butyrophilin like 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308245.1 | Intron | NP_001295174.1 | ||||
NM_152547.4 | Intron | NP_689760.2 | ||||
XM_006714823.3 | Intron | XP_006714886.1 | ||||
XM_006714824.3 | Intron | XP_006714887.1 | ||||
XM_006714825.3 | Intron | XP_006714888.1 | ||||
XM_011534444.2 | Intron | XP_011532746.1 | ||||
XM_017009113.1 | Intron | XP_016864602.1 | ||||
XM_017009114.1 | Intron | XP_016864603.1 | ||||
XM_017009115.1 | Intron | XP_016864604.1 | ||||
XM_017009116.1 | Intron | XP_016864605.1 | ||||
XM_017009117.1 | Intron | XP_016864606.1 |
MIR8089 - microRNA 8089 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |