Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTGAAAATCTGGAGGAAGAGAC[A/G]GAAAACATAATTGCAGCAAATCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604507 | ||||||||||||||||||||
Literature Links: |
BRD9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRD9 - bromodomain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIP13 - thyroid hormone receptor interactor 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166260.1 | 493 | Silent Mutation | ACA,ACG | T,T 116 | NP_001159732.1 | |
NM_004237.3 | 493 | Silent Mutation | ACA,ACG | T,T 116 | NP_004228.1 | |
XM_011514163.2 | 493 | Silent Mutation | ACA,ACG | T,T 116 | XP_011512465.1 |