Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGCCCAGAGTTTGCTGTTGAG[A/G]ATCAGCCTTAGACTGCCTTGATTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616391 | ||||||||||||||||||||
Literature Links: |
RANBP3L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RANBP3L - RAN binding protein 3 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161429.2 | 1482 | Silent Mutation | ATC,ATT | I,I 377 | NP_001154901.1 | |
NM_001323273.1 | 1482 | Silent Mutation | ATC,ATT | I,I 352 | NP_001310202.1 | |
NM_001323274.1 | 1482 | Silent Mutation | ATC,ATT | I,I 377 | NP_001310203.1 | |
NM_001323275.1 | 1482 | Silent Mutation | ATC,ATT | I,I 357 | NP_001310204.1 | |
NM_001323276.1 | 1482 | Silent Mutation | ATC,ATT | I,I 357 | NP_001310205.1 | |
NM_001323277.1 | 1482 | Silent Mutation | ATC,ATT | I,I 352 | NP_001310206.1 | |
NM_001323278.1 | 1482 | Silent Mutation | ATC,ATT | I,I 332 | NP_001310207.1 | |
NM_001323279.1 | 1482 | Silent Mutation | ATC,ATT | I,I 328 | NP_001310208.1 | |
NM_001323280.1 | 1482 | Silent Mutation | ATC,ATT | I,I 332 | NP_001310209.1 | |
NM_145000.4 | 1482 | Silent Mutation | ATC,ATT | I,I 352 | NP_659437.3 | |
XM_006714452.3 | 1482 | Silent Mutation | ATC,ATT | I,I 377 | XP_006714515.1 | |
XM_006714453.3 | 1482 | Silent Mutation | ATC,ATT | I,I 357 | XP_006714516.1 | |
XM_006714455.3 | 1482 | Silent Mutation | ATC,ATT | I,I 332 | XP_006714518.1 |