Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCTCCATCTCTGGAGTTCTTGC[A/G]TGGCATTTCCACCAGGGGGAGCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606323 | ||||||||||||||||||||
Literature Links: |
CYFIP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYFIP2 - cytoplasmic FMR1 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037333.2 | 741 | Intron | NP_001032410.1 | |||
NM_001291721.1 | 741 | Intron | NP_001278650.1 | |||
NM_001291722.1 | 741 | Intron | NP_001278651.1 | |||
NM_014376.3 | 741 | Intron | NP_055191.2 | |||
XM_011534516.2 | 741 | Intron | XP_011532818.1 | |||
XM_017009341.1 | 741 | Intron | XP_016864830.1 | |||
XM_017009342.1 | 741 | Intron | XP_016864831.1 |
FNDC9 - fibronectin type III domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001343.3 | 741 | Missense Mutation | CGC,TGC | R,C 185 | NP_001001343.2 |